Ashcroft D430IWD 0 to 30" H2O Differential General-Purpose Watertight Switch
Representative image only.
Ashcroft Mfr # D4-24-B-XCY-30IWD – Item # EW-68050-20
Trigger alarms or shut down equipment when pressure rises to setpoint
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Pressure (in WC)0
  • Max Pressure (in WC)30
  • Min Temperature (° F)0
  • Max Temperature (° F)150
  • Min Temperature (° C)-18
  • Max Temperature (° C)65
  • Wetted MaterialsBuna N, epoxy-coated carbon steel
  • Process Connection1/4\" NPT(F)
  • Unit Of Measurement\" WC
  • Electrical Connections3/4\" NPT(F) conduit
  • Switch output15 A at 125/250/480 VAC; 0.5 A at 125 VDC; 0.25 A at 250 VDC
  • OutputSPDT
  • Enclosure RatingNEMA 3, 4, 4X, 13; IP66
  • Media CompatibilityGases and liquids compatible with wetted materials
  • Description0 to 30\" H2O Differential General-Purpose Watertight Switch
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards AssociationUL Listed.
Key Features
  • 1 year warranty
More About this Item
General-Purpose Watertight Dual Set Point Switches are ideal for use with pumps, paint spraying equipment, liquid level, and presses. Use these watertight switches in breweries, dairies, chemical plants, and pulp/paper mills.
Housing meets NEMA 4X standards. Epoxy-coated aluminum enclosure mounts with integral wall mounting bracket with two additional knock-out plugs for installation. SS internal parts. Remove cover to read scale while adjusting set points.