Ashcroft LTA Temperature Switch Remote Mount, 15' Capillary, 65/125°C, Adjustable Deadband w/ Adapter
Representative image only.
Ashcroft Mfr # LTA N4 H 15-A7-030 XBX 65/125C – Item # EW-39011-54
Prevent unneeded activation with adjustable deadband
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—meet NEMA 4, 4X, and IP66 requirements
PRODUCT OPTIONS
$701.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Process Connection1/2" NPTM Adapter
  • Electrical Connections(1) 3/4" NPTF
  • Min Temperature (° C)65
  • Temperature Accuracy±1%
  • Height (in)21 9/16
  • Width (in)4
  • DescriptionTemperature Switch Remote Mount, 15' Capillary, 65/125°C, Adjustable Deadband w/ Adapter
Key Features
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—meet NEMA 4, 4X, and IP66 requirements
More About this Item
Single set point, adjustable deadband, SPDT switches feature epoxy-coated aluminum systems for reliable performance. Choose direct-mounting units with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration, velocity, or pressure. Remote-mount units have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.