Ashcroft Pressure Switch 0 to 30 PSI; 1/4" NPT(F)

Representative image only.
Ashcroft Mfr # FPSN7PV07 30PSI – Item # EW-68350-68
Approved for Class I and II hazardous indoor locations
  • Enclosure is NEMA 7/9 and IP66 rated
  • Watertight anodized aluminum body for environmental protection
  • Diaphragm-sealed piston actuator offers long, reliable service
  • Hermetically sealed snap-acting switch element
  • Field adjustable
  • Factory sealed leads
  • Setpoint adjustable from 15-100% of range
  • Simple to install and use
  • Class I, Groups A,B,C, or D and Class II, Group E,F, or G
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Accuracy±1% full-scale
  • Min Pressure (PSI)0
  • Max Pressure (PSI)30
  • Min Temperature (° F)20
  • Max Temperature (° F)200
  • Min Temperature (° C)-7
  • Max Temperature (° C)93
  • Wetted Materials316 SS, PTFE, Viton®
  • CaseAluminum
  • Process Connection1/2\" NPT(F)
  • Battery Type125/250 VAC, 3-wire
  • Electrical Connections1/2\" conduit
  • Output5 A at 125/250 VAC
  • Enclosure RatingNEMA 7/9, IP66
  • Media CompatibilityGases and liquids compatible with wetted materials
  • DescriptionPressure Switch 0 to 30 PSI; 1/4\" NPT(F)
  • Warranty1 year
1 Year WarrantyConformite EuropeenneCanadian Standards AssociationIntrinsically SafeIP 66 RatedUL Listed.

Key Features

  • Enclosure is NEMA 7/9 and IP66 rated
  • Watertight anodized aluminum body for environmental protection
  • Diaphragm-sealed piston actuator offers long, reliable service
  • Hermetically sealed snap-acting switch element
  • Field adjustable
  • Factory sealed leads
  • Setpoint adjustable from 15-100% of range
  • Simple to install and use
  • Class I, Groups A,B,C, or D and Class II, Group E,F, or G
  • 1 year warranty

More About this Item

These Ashcroft F-Series pressure switches are designed to satisfy most requirements for alarm, shutdown, control and interlock applications on a wide variety of process and equipment. Electromechanical pressure switches are usually more cost effective than transmitters for these types of applications. Materials of construction have been selected for long life.

Applications
Hazardous locations
Engines and compressors
Corrosive environments
Process applications
Offshore applications
Pipelines
Machine tools
Replacement and retrofit
Where size is a consideration or equipment is being downsized

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat