Ashcroft T7 Temperature Switch Direct Mount 6.0" Bulb 110/190C
Representative image only.
Ashcroft Mfr # T7 24 TS-060 110/190C – Item # EW-39012-91
Explosion proof and waterproof for your toughest jobs
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—switches meet Division 1 and 2, NEMA 7 and 9, and IP66 requirements
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Switch Direct Mount 6.0\" Bulb 110/190C
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—switches meet Division 1 and 2, NEMA 7 and 9, and IP66 requirements
  • 1 year warranty
More About this Item
Single set point, fixed deadband, SPDT switches feature 316 stainless steel systems for long-lasting performance—even in harsh environments. Choose direct-mounting units with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration or pressure. Remote-mount switches have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.