Ashcroft T7 Temperature Switch Remote Mount 5' CAPIllary 235/375F with Adapter
Representative image only.
Ashcroft Mfr # T7 24 T05-030 XBX 235/375F – Item # EW-39013-03
Explosion proof and waterproof for your toughest jobs
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—switches meet Division 1 and 2, NEMA 7 and 9, and IP66 requirements
PRODUCT OPTIONS
$720.75USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Switch Remote Mount 5' CAPIllary 235/375F with Adapter
Key Features
  • Quick, accurate response via SAMA Class II vapor pressure thermal system
  • Choose the switch to meet your needs—wide variety of lengths and temperature ranges available
  • Easy to operate and install—switches meet Division 1 and 2, NEMA 7 and 9, and IP66 requirements
More About this Item
Single set point, fixed deadband, SPDT switches feature 316 stainless steel systems for long-lasting performance—even in harsh environments. Choose direct-mounting units with rigid probes—great for use with 3/8" nominal bore thermowells in areas with high vibration or pressure. Remote-mount switches have spring-mounted capillaries with 302 stainless steel armor and include a 1/2" NPT(M) bushing adapter.