Ashcroft XLDP0.50"WC Differential Pressure Transmitter, 0.50" WC
Representative image only.
Ashcroft Mfr # XL-3-MB2-42-ST-P5IW – Item # EW-68439-18
  • FEATURES
  • 15 psid proof pressure for all pressure ranges
  • 0.25% accuracy is standard (.50% FS accuracy available)
  • 4-20mA output is standard (1-5V or 1-6V DC outputs available)
  • A wide assortment of uni-directional and bi-directional ranges available
  • Non standard ranges available
  • All stainless steel NEMA 2 construction
  • 1/4" barbed stainless steel connection is standard
  • Factory adjustable damping from 5msec to 1 sec
  • On board voltage regulation allows use of lower cost, unregulated power supply
  • Three year warranty
  • CE approved
  • 3 year warranty
  • SPECS & DESCRIPTION
    Specifications & Description
    • Accuracy±0.25% full-scale
    • Min Pressure (in WC)0
    • Max Pressure (in WC)0.5
    • Process Connection1/4\" NPT(F)
    • Output4 to 20 mA
    • DescriptionDifferential Pressure Transmitter, 0.50\" WC
    • Warranty3 year
    3 Year Warranty
    Key Features
    • FEATURES
    • 15 psid proof pressure for all pressure ranges
    • 0.25% accuracy is standard (.50% FS accuracy available)
    • 4-20mA output is standard (1-5V or 1-6V DC outputs available)
    • A wide assortment of uni-directional and bi-directional ranges available
    • Non standard ranges available
    • All stainless steel NEMA 2 construction
    • 1/4" barbed stainless steel connection is standard
    • Factory adjustable damping from 5msec to 1 sec
    • On board voltage regulation allows use of lower cost, unregulated power supply
    • Three year warranty
    • CE approved
  • 3 year warranty
  • More About this Item
    This differential transmitter is a highly reliable variable capacitance sensor using a SiGlas silicon and glass construction. This ultra-thin single crystal diaphragm is specifically designed to detect less than one micron of diaphragm deflection