All Test Pro Condition Calculator 4.1 Software
Representative image only.
All Test Pro Mfr # AT3402-4 – Item # EW-20006-92
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCondition Calculator 4.1 Software
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Find the motor faults you miss with a megohmmeter or PdM.
APPLICATIONS
  • Test new and rewound motors before installing to insure they’re good. dangerous leaks
  • Test motors during shut-down to detect developing faults
  • Positively troubleshoot motor and drive trips to identify the problem area – motor, drive, overload, cabling, etc.
  • When testing installed motors, most tests can be run from the MCC through even hundreds of feet of cable
  • Detects all of the following:
    • Turn-to-turn faults
    • Coil-to-coil and phase-to-phase faults
    • Open phases
    • Phase unbalances
    • Broken rotor bars
    • Grounded windings
    • Capacitor failures (pass/fail test)
    • Shorted windings
  • Works on all sizes of motors—fractional through thousands of HP
  • Low-voltage tests are non-destructive