Atago MASTER-SUR/NM Refractometer; 1.000-1.060 Urine SG/0.0-12.0 Serum Protein
Representative image only.
Atago Mfr # 2773 MASTER-SUR/NM – Item # UX-02942-09
Measure serum protein, specific gravity of urine, and refractive index – with one instrument!
  • Rugged, high impact plastic design that is corrosion resistant
  • 1 year warranty
PRODUCT OPTIONS
$483.25USD / Each
$121.50 - $195.50USD / Each
for an additional
$121.50USD / Each
for an additional
$195.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Calibration Standard Solution Sets
Refractive Index Standard Solutions
$483.75 - $1,508.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Resolution0.001 (s.g. urine); 0.2 (g serum protein); 0.0005 (RI)
  • Accuracy±0.001 (s.g. urine); ±0.2 (g serum protein); ±0.0005 (RI)
  • DescriptionRefractometer; 1.000-1.060 Urine SG/0.0-12.0 Serum Protein
  • Warranty1 year
1 Year Warranty
Key Features
  • Rugged, high impact plastic design that is corrosion resistant
  • 1 year warranty
More About this Item
Use these refractometers to provide precise and accurate readings for urine specific gravity, serum protein, and refractive index. The large easy-to-read display is screen shaded and the rubber eye guard allows you to comfortably take your measurements. Read your measurements in seconds after placing only a few drops of sample on the prism.