Atago MASTER-URC/NM Refractometer, 1.000 to 1.050 urine SG, 1.333 to 1.356 nD refractive index
Representative image only.
Atago Mfr # 2793 MASTER-URC/NM – Item # EW-02942-05
Measure serum protein, specific gravity of urine, and refractive index – with one instrument!
  • Rugged, high impact plastic design that is corrosion resistant
  • 1 year warranty
PRODUCT OPTIONS
$378.25USD / Each
$115.50 - $188.00USD / Each
for an additional
$115.50USD / Each
for an additional
$188.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
Calibration Standard Solution Sets
Refractive Index Standard Solutions
$465.00 - $1,450.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Resolution0.001 (s.g. urine); 0.001 (nD)
  • Accuracy±0.001 (s.g. urine); ±0.001 (nD)
  • DescriptionRefractometer, 1.000 to 1.050 urine SG, 1.333 to 1.356 nD refractive index
  • Warranty1 year
1 Year Warranty
Key Features
  • Rugged, high impact plastic design that is corrosion resistant
  • 1 year warranty
More About this Item
Use these refractometers to provide precise and accurate readings for urine specific gravity, serum protein, and refractive index. The large easy-to-read display is screen shaded and the rubber eye guard allows you to comfortably take your measurements. Read your measurements in seconds after placing only a few drops of sample on the prism.