Atago RE-6705 Sample Tube for Semiautomatic Polarimeter, 100 mm, 5 mL
Representative image only.
Atago Mfr # RE-6720 – Item # EW-81200-50
Extend the life of your Atago Polarimeter with these replacement tubes
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionSample Tube for Semiautomatic Polarimeter, 100 mm, 5 mL
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Replacement tube 81200-21 is suitable for use with the 81200-15 Atago Polarimeter. Tube models 81200-50 and -52 are compatible with Polarimeter 81200-00.
Use the 100-mm tube for cloudy, impurity-filled samples.