Autovial AV125UAQU Whatman Syringeless Filters, PVDF; 0.45µ; 50/box
Representative image only.
Autovial Mfr # AV125UAQU – Item # EW-29703-22
Direct instrument injection
  • Use PVDF filters (<60% organic) with low nonspecific protein binding for samples in aqueous solution and solvent or aqueous/organic solvents
PRODUCT OPTIONS
$294.00USD / of 50
SPECS & DESCRIPTION
Specifications & Description
  • Membrane MaterialPolyvinylidene fluoride (PVDF)
  • Pore Size (µm)0.45
  • Housing MaterialPolypropylene (PP)
  • DescriptionWhatman Syringeless Filters, PVDF; 0.45µ; 50/box
Key Features
  • Use PVDF filters (<60% organic) with low nonspecific protein binding for samples in aqueous solution and solvent or aqueous/organic solvents
More About this Item
Autovial syringeless filters offer maximum convenience. Simply fill the 12-mL graduated polypropylene filter barrel and press the plunger to push sample through. Features integral filter, built-in air purge, and recessed slip-luer tip (needle may be attached for direct instrument injection).
Select your filter based on membrane material and its compatibility to your sample (see application selection guide below). Membrane materials are nylon, PTFE (PTFE), polyvinylidene fluoride (PVDF), glass microfiber (GMF), and mixed ester (ME).