Aven Tools Titanium Precision Smooth Fine Tip 3 Tweezers, 4.75" L; 1/Pk
Representative image only.
Aven Tools Mfr # 18053TT – Item # EW-07289-28
$45.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • MaterialTitanium
  • SterileNo
  • DescriptionTitanium Precision Smooth Fine Tip 3 Tweezers, 4.75" L; 1/Pk
More About this Item
Titanium tweezers are excellent for the silicon chip industry, for handling microscopic particles, and when working in a magnetic field as they are non-magnetic, lighweight, and strong. They do not shed particles like stainless steel tweezers and are biocompatible. Temperature resistance is up to 800°F(426°C).