AW-Lake FEM-03 Gear Flow Sensor/Transmitter Accessory, Flow Monitor
Representative image only.
AW-Lake Mfr # MX9-SX-XX – Item # EW-32928-75
$714.00USD / Each
$310.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionGear Flow Sensor/Transmitter Accessory, Flow Monitor
More About this Item
Monitor both flow rate and flow from pulse output flowmeters in any engineering unit with a push of a button. Programmable, compact, and easy-to-use. Input square wave, sine, or sawtooth frequency. Back-up totalizer lock-out feature prevents accidental erasure. High-speed microprocessor ensures fast and accurate reporting — includes a 4 to 20 mA output for recording or datalogging. Model 32928-76 has two channels to monitor dual flows and display them separately, as a sum, as a difference, or as a ratio.