B&K Precision 1211E Handheld NTSC Generator
Representative image only.
B&K Precision Mfr # 1211E – Item # EW-20017-09
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Max Frequency Range (MHz)61.25
  • DescriptionHandheld NTSC Generator
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This handheld NTSC pattern generator is powered by a 9 volt battery or AC adapter providing NTSC color bars, crosshatch, dot, staircase, circle, center cross, windows and a full range of color raster patterns. The outputs available are composite video via RCA connector, a RF output BNC connector, or an S-Video mini-DIN output. Signals can be produced as an interlaced or progressive scanning mode.
FEATURES
  • S-Video, composite video, audio output
  • Interlace or progressive scanning system
  • 9 VDC battery (included)
  • AC adapter available