This item is now discontinued. We recommend the following as suggested replacements: EW-26868-18
B&K Precision 1710A Analog DC Power Supply 0-30V, 0-1A
Representative image only.
B&K Precision Mfr # 1710A – Item # EW-20017-13
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Output Voltage0 to 30 VDC
  • Output Current0 to 1 A
  • DescriptionAnalog DC Power Supply 0-30V, 0-1A
  • Warranty1 Year
1 Year WarrantyConformite Europeenne
Key Features
  • 1 year warranty
More About this Item
FEATURES
  • Largest selection of voltage and current ratings ever offered
  • Connect two supplies in parallel to double the current output
  • Connect two supplies in series to double the voltage output
  • Reliable, Durable
  • Operate continuously at full load without overheating
  • Fully overload protected
  • Coarse and fine voltage controls
  • Excellent regulation
  • Very low ripple
  • Constant voltage or constant current operation
  • Continuously monitor voltage and current output on two meters