B&K Precision 391A Portable Test Bench True RMS DMM with Rubberized Case
Representative image only.
B&K Precision Mfr # 391A – Item # EW-20005-15
$260.00USD / Each
$81.50 - $119.00USD / Each
for an additional
$119.00USD / Each
for an additional
$81.50USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • AC Max (amps)391
  • Safety RatingCAT III 1000V
  • DescriptionPortable Test Bench True RMS DMM with Rubberized Case
More About this Item
FEATURES
  • High quality, ruggedized, multifunction
  • Resistance measurement
  • Diode check
  • Frequency measurement
  • Audible continuity
  • All current ranges fused
  • Ruggedized case
  • Auto power off
  • Designed to meet IEC61010-1 CATIII 1000V, class 2
Call Us At1-847-549-7600
Live chat