B&K Precision 844USB EPROM Device Programmer
Representative image only.
B&K Precision Mfr # 844USB – Item # EW-20020-91
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEPROM Device Programmer
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
BP352125
  • Small, fast and powerful programmer
  • 40 pin DIP ZIF socket, no adapter required for device in DIP package
    up to 40 pins
  • In-circuit serial programming (ISP) capability included
  • High-speed IEEE 1284 printer-port interface, up to 1MB/sec. Transfer rate
BP352126
  • EPROM, EEPROM, Flash, NVRAM and serial EEPROM capability
  • Low Cost
  • 32 pin ZIF socket, no adapter required for any DIP device up to 32pins
  • Parallel printer port interface
BP35212
  • Fast and accurate programming
  • 48 pin ZIF socket, no adapter required for any DIP device up to 48 pins
  • USB and Parallel printer interface
  • Thousands of supported devices
BP352123
  • Fast and accurate programming
  • 48 pin ZIF socket, no adapter required for any DIP device up to 48
    pins
  • Parallel printer port interface
  • Gang programming capability