B&K Precision 851 Deluxe Heavy-Duty EPROM Eraser
Representative image only.
B&K Precision Mfr # 851 – Item # EW-20020-82
$195.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionDeluxe Heavy-Duty EPROM Eraser
More About this Item
BP447317 This is a heavy duty EPROM eraser that can simultane-ously erase up to 40 twenty-four pin EPROM’s. It is con-structed of an all-metal case plus an advanced designedchip drawer that prevents UV radiation from being a haz-ard to the user. The chip drawer is lined with conductivefoam to prevent electrostatic damage to the chips. BP447318This is a UV-EPROM eraser featuring small dimensions,high erasing efficiency, and simple operation. Up to fiveEPROM’s in 40-pin DIP packages can be erased simultane-ously. A built-in timer is preset to an optimum erasing peri-od.EPROM
  • Indicator light when erasing
  • Power safety switch inside the drawer shuts off unit whenever thedrawer is opened
  • Large capacity
  • One year warranty