Bacharach 0019-7051 Combustible Gas Leak Detector

Representative image only.
Bacharach Mfr # H0019-7051 – Item # EW-10103-94
30-hour battery life for extended field use
  • Ten bright red LEDs and audible tick indicate relative gas concentration
  • Turn thumbwheel to power on and adjust tick rate for easy one-handed operation
  • Extra-long 20" (50.8 cm) flexible probe for hard-to-reach places
  • UL 913 classified as intrinsically safe in Class I, Division 1, Groups A, B, C, and D hazardous locations

SPECS & DESCRIPTION

Specifications & Description

  • Gas MeasuredCombustible Gases
  • Methane Sensitivity20 ppm
  • Probe20\" (50.8 cm) gooseneck
  • ApprovalUL913 intrinsically safe for Class 1, Div 1, Groups A, B, C, and D
  • Battery TypeFive C batteries (not included)
  • Battery Life30 hours typical
  • Length (in)8 1/2
  • Width (in)2 1/4
  • Depth (in)1 3/4
  • Length (cm)21.59
  • Width (cm)5.7
  • Depth (cm)4.4
  • DescriptionCombustible Gas Leak Detector

Key Features

  • Ten bright red LEDs and audible tick indicate relative gas concentration
  • Turn thumbwheel to power on and adjust tick rate for easy one-handed operation
  • Extra-long 20" (50.8 cm) flexible probe for hard-to-reach places
  • UL 913 classified as intrinsically safe in Class I, Division 1, Groups A, B, C, and D hazardous locations

More About this Item

Applications include: acetone, acetylene, ammonia, benzene, butane, ethanol, gasoline, hexane, hydrogen, industrial solvents, methane, naptha, natural gas, paint thinners, and propane.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat