Bacharach 3015-4200 Refrigerant Monitor, Fixed, 120VAC, 60Hz
Representative image only.
Bacharach Mfr # 3015-4200 – Item # EW-10103-87
Non-dispersive infrared detector eliminates false alarms
  • Helps comply with federal, state, and local safety codes governing gas emissions
  • Maintain equipment efficiency, promote safety, and protect the environment
  • Continuously test areas up to 500 feet from the monitor
  • Use default alarm levels or adjust to more practical levels
  • Automatic purge uses ambient area to zero the monitor
$4,860.50USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionRefrigerant Monitor, Fixed, 120VAC, 60Hz
Key Features
  • Helps comply with federal, state, and local safety codes governing gas emissions
  • Maintain equipment efficiency, promote safety, and protect the environment
  • Continuously test areas up to 500 feet from the monitor
  • Use default alarm levels or adjust to more practical levels
  • Automatic purge uses ambient area to zero the monitor
  • Exhaust port allows connection of tubing to vent gases up to 100 feet away
  • Front panel lights indicate system is on, system fault, and alarm condition
  • Memory stores last 200 measurements and the last 30 alarm events with date and time stamp
More About this Item
The HGMSZ is designed for continuous single-point monitoring of CFC, HFC, HCFC, and Halon refrigerant gases. Turnkey monitors are easy to use: mount on a wall, hard wire, install tubing, select refrigerant and you’re ready to measure. Front panel LCD shows the type of gas being monitored, current gas level, and peak gas level. The monitor retains a log of previous readings that can be easily accessed for analysis. Set the logging interval from 1 to 9999 minutes. Audible alarm and front panel indicators signal alarm and fault conditions. Relay contacts can be used to trigger external alarm devices in the event of a system fault, or if a leak, spill, or evacuation level of gas is detected. The system also includes a 4-20 mA output that can be connected to remote monitoring equipment.