Dunlop 86049 SIZE 11 Hip Boots, 28 inch Height, US Mens 11 | Womens 13 Shoe; 1 Pair
Representative image only.
Dunlop Mfr # 86049 SIZE 11 – Item # EW-86420-11
Stay warm and dry out in the field
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)28
  • Men's Size11
  • Womens Size13
  • DescriptionHip Boots, 28 inch Height, Mens US Size 11 Shoe; 1 Pair
More About this Item
Durable PVC exterior resists oil, grease, gasoline, saltwater, and ozone but maintains flexibility even in cold weather. Lightweight, nonabsorbent polyester lining allows easy cleaning. Hip boots and chest waders feature steel toes for impact protection, and a cleated outsole for excellent traction.
Wear over-the-sock.
Made in the USA.
Steel toe complies with ASTM F2413-11
Hip boots are 28" length to provide protection above the knee. Adjustable nylon strap keeps boots in place.
Chest waders are 54" long with top dawstring closure for a snug fit at the chest. Nylon, H-back suspenders with reinforced buttons keep waders in place. Built-in belt loop with adjustable belt for added security.