Bausch & Lomb Hastings Triplet Magnifier, 10X, 0.62" Diameter
Representative image only.
Bausch & Lomb Mfr # 81-61-71 – Item # EW-03882-22
Provides sharp, distinct images without distortion
  • Lenses are recessed to protect it from damage
  • Swing-away, nickel-plated case also protects the lens and acts as a handle
  • 1 year warranty
$86.70USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Magnification10X
  • Viewing Area0.62\" (15.8 mm) diameter
  • LensTriple
  • Focal Length1\" (25.4 mm)
  • DescriptionHastings Triplet Magnifier, 10X, 0.62\" Diameter
  • Warranty1 Year
1 Year Warranty
Includes
protective storage pouch.
Key Features
  • Lenses are recessed to protect it from damage
  • Swing-away, nickel-plated case also protects the lens and acts as a handle
  • 1 year warranty
More About this Item
These magnifiers eliminate the distortion commonly found with similarly priced pocket magnifiers by bonding together three optical lenses, forming a "Hastings Triplet". Hastings Triplet Magnifiers incorporate three separate glass lenses bonded together to form a compound lens to provide sharp, very distinct magnified images without distortion.