This item is now discontinued. We recommend the following as suggested replacements: EW-07944-28
BD Biocoat 136898 60 mL plastic disposable syringe, luer-lock tip, 40 / box
Representative image only.
BD Biocoat Mfr # 136898 – Item # EW-07940-30
  • Ideal for use with syringe filters
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeLuer Lock
  • Syringe Capacity (cc)60
  • Syringe Capacity (mL)60
  • Graduations (mL)1
  • Graduations (cc)1
  • MaterialPolypropylene
  • SterileYes
  • Description60 mL plastic disposable syringe, luer-lock tip, 40 / box
Key Features
  • Ideal for use with syringe filters
More About this Item
Use these disposable, sterile, polypropylene syringes for general-purpose applications. Syringes come in an easy-to-open sealed package and can be autoclaved out of the package. Needles are not included; order separately.
individually wrapped, sterile