B-D Disposable Syringe, Non-Sterile, Slip-Tip, Bulk Pack, 30 mL; 225/Cs
Representative image only.
B-D Mfr # 301034 – Item # EW-07945-22
Male Luer-Lok™ or slip tips allow for easy connection of syringe filters
  • Constructed of durable polypropylene
  • Autoclavable
  • 30 mL capacity
  • Slip-tip connection
  • Available in bulk packages
PRODUCT OPTIONS
$264.00USD / Case of 225
SPECS & DESCRIPTION
Specifications & Description
  • Syringe TypeSlip Tip
  • Syringe Capacity (cc)30
  • Syringe Capacity (mL)30
  • Graduations (mL)1
  • Graduations (cc)1
  • MaterialPolypropylene
  • DescriptionDisposable Syringe, Non-Sterile, Slip-Tip, Bulk Pack, 30 mL; 225/Cs
Key Features
  • Constructed of durable polypropylene
  • Autoclavable
  • 30 mL capacity
  • Slip-tip connection
  • Available in bulk packages
More About this Item
Use these latex-free, disposable polypropylene syringes for your general-purpose applications. Available with either Luer-Lok or slip-tip connections. Non-sterile syringes are available in standard and bulk package options. Needles and shield tips are not included, order separately.