Bead bag for Waterless Ice Baths or Buckets
Representative image only.
Lab Armor Item # EW-06274-37
Replace water in laboratory water baths and ice in ice buckets.
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Easily tote beads to and from the freezer
  • 1 year warranty
PRODUCT OPTIONS
$52.30USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBead bag for waterless ice baths or buckets
  • Warranty1 year
1 Year Warranty
Key Features
  • Reduce the risk of contamination, achieve reproducible results and have greater confidence in your work
  • No need to worry about burnout as there is no water to evaporate.
  • Highly recyclable, fewer harmful biocides, and more energy efficient beads are eco-friendly
  • Easily tote beads to and from the freezer
  • 1 year warranty
More About this Item
Compatible with waterless ice buckets, items 06274-24 and -28.
Perfect companion to tote the beads to and from the freezer. It’s the nylon mesh that allows any condensation on the beads to escape. The nylon fiber also resists mold and mildew. Should Beads need to washed, the mesh bag works like a wash bag. The mesh holes are small enough to hold the beads in and makes washing the beads easy.