Beijer Electronics iX Series Rugged HMI, 15" Extra-Bright Glossy, High Performance, CAN, 12-24 VDC
Representative image only.
Beijer Electronics Mfr # IXT15BR/4525-640004801 – Item # EW-45004-25
Operate in extreme temperatures and other harsh or dangerous environments
  • Extensive certifications ensure function despite heat, cold, water, vibration, or shock
  • Galvanic isolation protects against both bursts and surges in electricity
  • IP66/NEMA 4X front-panel enclosure resists snow, rain, high-pressure washdown, dirt, and dust
  • Vibration/shock: 4 g RMS sweep sine (vibration), 40 g 11 ms half-sine (shock)
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Communication OutputTwo Ethernet (1 x 10/100 Base-T, 1 x 10/100/1000 Base-T), two serial (1 x RS-232, 1 x RS-422/485), three USB (2.0 host), CAN module
  • Memory CapacityRAM: 1 GB DDR2; Flash: 4 GB SLC NAND; External storage media: one SD card slot
  • Enclosure RatingIP66, NEMA 4X (front), IP20 (rear)
  • Display TypeWidescreen TFT LCD with analog-resistive touch screen and LED backlight
  • Power (VDC)12 or 24
  • DescriptionRugged HMI, 15\" Extra-Bright Glossy, High Performance, CAN, 12-24 VDC
  • Warranty2 Years
Two year warranty
Key Features
  • Extensive certifications ensure function despite heat, cold, water, vibration, or shock
  • Galvanic isolation protects against both bursts and surges in electricity
  • IP66/NEMA 4X front-panel enclosure resists snow, rain, high-pressure washdown, dirt, and dust
  • Vibration/shock: 4 g RMS sweep sine (vibration), 40 g 11 ms half-sine (shock)
  • 2 year warranty
More About this Item
Ideally suited to oil and gas, marine, and water/wastewater industries, and solar array or engine room controls, these human machine interfaces (HMI) feature specialized components to increase durability and undergo rigorous testing to ensure operation under the toughest conditions. Hazardous environment certifications include UL Class I Div 2, ATEX Zone 2, and IECEx Zone 2. Marine environment certifications include DNV, ABS, GL, LR, KR, and EN60945.
Both the serial and the CAN ports are galvanically isolated, embedded NAND flash memory is designed for extreme temperatures and extended write cycles, and touch screen is composed of chemically strengthened glass for longer service life. The panels accept 12 or 24 VDC, so converters are unnecessary.
Optional features:
High-performance models (45004-22, -23, -25, -26) feature an enhanced processor. The standard is 1.0 GHz, while the high performance version is 1.6 GHz.
High-brightness models (45004-22, -23, -25, -26, -28, -29) offer a brighter backlight suitable for easy reading in direct sunlight. Both the standard and the high-brightness models are dimmable for low-light conditions.
CAN module (45004-24 to -26, -29) allows HMI to interface with controllers using CAN communication channels.
Glossy display (45004-22 and -25) is best suited for panels installed in low-light or diffuse-light conditions. The reflective coating provides better contrast and color saturation than the standard matte coating; however, it also suffers from glare in bright rooms and on sunny days.