Bel-Art 18206 Double -Style Burette Clamp
Representative image only.
Bel-Art Mfr # 182060000 – Item # EW-06703-12
$79.50USD / Each
BUY MORE & SAVE
  • 1-5$79.50USD / Each
  • 6-29$71.55USD / Each
  • 30-119$67.58USD / Each
  • 120+$63.60USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialPolypropylene (PP)
  • Connector MaterialPolypropylene (PP)
  • Accepts Rod Diameter (in)0.375
  • DescriptionDouble -Style Burette Clamp
More About this Item
Polypropylene (PP) clamps hold burettes without covering graduations. Clamps include 3/8" and 1/2" bushings to fit any standard support rod. Stand includes 5-1/2 lb weighted base and 30 (76.2 cm) L x 1/2" (1.3 cm) dia rod. Base and rod are also PP.