Bel-Art Autoclavable HDPE Biohazard Bags, 10 to 12 Gallon, 25 x 35"; 200/CS

Representative image only.
Bel-Art Mfr # F13164-2535 – Item # EW-06494-04
Hold more—up to 50 gallons
  • No misinterpretations—procedures printed in four languages
  • Know when it's autoclaved—indicator
  • patch darkens for verification
  • Autoclavable at 250°F (121°C).

PRODUCT OPTIONS

$402.00USD / Case of 200

SPECS & DESCRIPTION

Specifications & Description

  • ColorOrange and Red
  • MaterialHDPE
  • Capacity (Gal)12
  • Length (in)35
  • Width (in)25
  • Length (cm)88.9
  • Width (cm)63.5
  • Thickness (mil)1
  • Description10 to 12 Gallon Autoclavable HDPE Biohazard Bags, 25 x 35"; 200/cs

Key Features

  • No misinterpretations—procedures printed in four languages
  • Know when it's autoclaved—indicator
  • patch darkens for verification
  • Autoclavable at 250°F (121°C).

More About this Item

Bright orange-red, 1.25 mil, high-molecular weight HDPE bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat