Bel-Art F12416-0019 GHS Labeled Safety-Vented Ethanol Wash Bottles, LDPE, 500 mL; 4/Pk
Representative image only.
Bel-Art Mfr # F12416-0019 – Item # EW-62300-73
Eliminate spout dripping with built-in safety vent
  • Quick identification of contents with color-coded caps and chemical identification label
  • Fill bottles easily with wide-mouth design
  • Compliant with OSHA regulations
  • Excellent chemical compatibility with LDPE bottle and polypropylene (PP) cap
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (mL)500
  • Capacity (oz)16
  • MaterialLow Density Polyethyene (LDPE)
  • Cap MaterialPolypropylene
  • Cap ColorNatural
  • Bottle ColorTranslucent
  • AutoclavableNo
  • Label TypeEthanol
  • DescriptionGHS Labeled Safety-Vented Ethanol Wash Bottles, PE, 500 mL; 4/Pk
  • Warranty3 month
3 Month Warranty
Key Features
  • Quick identification of contents with color-coded caps and chemical identification label
  • Fill bottles easily with wide-mouth design
  • Compliant with OSHA regulations
  • Excellent chemical compatibility with LDPE bottle and polypropylene (PP) cap
  • 3 month warranty
More About this Item
These safety-vented wash bottles provide compliance with the Globally Harmonized System (GHS) of Classification and Labeling of Chemicals and OSHA’s HazCom 2012 requirements. These wash bottles are pre-printed with the chemical name and formula, GHS pictogram and signal word, National Fire Protection Association (NFPA) Section 704 four-color diamond, health hazards, Chemical Abstract System (CAS) number, and suggested protective clothing and equipment. Bottles with labels that do not indicate a hazardous chemical do not have/require GHS pictogram or signal word. These bottles are Deionized Water, Distilled Water, Isotonic Saline, and Water.
These wash bottles feature a unique venting valve which prevents pressure build-up, eliminating potentially dangerous chemical drips. This feature also allows dispensing of the wash bottle in an upright or inverted position. The methyl ethyl ketone and toluene do not include vented valve. Available in a 4-piece assortment pack which includes one each of Acetone, Isopropanol, Bleach, and Ethanol bottles.