Bel-Art F65000-0000 Cordless Microtube Homogenizer with Batteries
Representative image only.
Bel-Art Mfr # 651000000 – Item # EW-44444-70
Process precipitates, pellets, tissue, and cells directly in the microtube!
  • Don't let power cords get in your way—homogenizer unit is battery operated
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
  • 1 year warranty
$116.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionCordless Microtube Homogenizer with Batteries
  • Warranty1 year
1 Year Warranty
Key Features
  • Don't let power cords get in your way—homogenizer unit is battery operated
  • Homogenize samples directly in their storage container
  • Obtain thorough mixtures with the tight pestle-to-microtube tolerance
  • Protect your samples—tubes and pestles are made from virgin polypropylene
  • 1 year warranty
More About this Item
The handheld, battery-operated homogenizer is lightweight and can handle a large quantity of samples for routine applications. Microtubes and pestles are made from animal and phthalate/phthalate-ester free polypropylene that is lot traceable and certified DNAse, RNAse, and pyrogen free. Tubes feature a snap-cap design that can be pierced and are frosted for easy labeling. Tube bottoms are also 25% thicker than traditional microtubes to maintain integrity during homogenization. Pestles are ribbed to provide a good grip during assembly and allow air to escape while mixing. Tubes and pestles are available as individually wrapped, bulk packed, or paired in tube/pestle sets.