Bel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
Representative image only.
Item # EW-06833-10
Deliver accurate regulation or interruption of fluid flow
  • Constructed of Delrin acetal
$35.00USD / Pkg of 3
SPECS & DESCRIPTION
Specifications & Description
  • Clamp MaterialDelrin acetal
  • Clamp TypeTubing
  • Max Grip Range (in)0.5
  • DescriptionBel-Art Flow Control Clamp, Delrin Screw Clamp, 3/Pk
Key Features
  • Constructed of Delrin acetal
More About this Item
DELRIN(R) resists weak acids, bases, and organic solvents. Controls flow in flexible tubing up to 1/2" (13 mm) OD. Replaces stopcocks in many applications. Install without disconnecting tubing. Autoclavable. Bel-Art.