Bel-Art Heavy-Duty Biohazard Disposal Bags, 25"W x 35"L

Representative image only.
Bel-Art Mfr # F13165-2535 – Item # EW-06495-04
Indicator patch darkens when autoclaved
  • Heavy-duty bag resists tears and leaks
  • Autoclavable at 300°F (149°C)

PRODUCT OPTIONS

$528.00USD / Pkg of 200

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (Gal)12
  • MaterialPolypropylene
  • ColorOrange and Red
  • Length (in)35
  • Width (in)25
  • Length (cm)88.9
  • Width (cm)63.5
  • Thickness (mil)2

Key Features

  • Heavy-duty bag resists tears and leaks
  • Autoclavable at 300°F (149°C)

More About this Item

These extra-strength, 2-mil thick bags are bright orange red. High-molecular weight polypropylene bags are printed with the biohazard warning symbol and precautionary procedures in four languages. The sterilization indicator patch darkens when autoclaved.

Optional bag stands are made of aluminum tubing with baked on epoxy coating. The stands fold flat for convenient storage and include a polystyrene safety drip tray that securely supports the bag in the stand. Order stands and plastic ties separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat