Bel-Art Paddle Flow Indicator, 0.5 to 10 L/min for 1/4" to 5/16" ID Tubing
Representative image only.
Bel-Art Mfr # 19937-0002 – Item # EW-06297-06
An inexpensive way to visually monitor flow of liquids and gases
  • Visually confirm flow with clear window
PRODUCT OPTIONS
$25.45USD / Pkg of 1
BUY MORE & SAVE
  • 1-11$25.45USD / Pkg of 1
  • 12+$22.91USD / Pkg of 1
$332.00USD / Each
Time to recalibrate? Click on the calibration service to view details, then order.
SPECS & DESCRIPTION
Specifications & Description
  • Min Flow Rate (LPM)0.5
  • Max Flow Rate (LPM)10
  • Max Operating Temperature (° F)185
  • Max Operating Temperature (° C)85
  • Pipe Size1/4" to 5/16" ID Tubing
  • Fluid Typeliquid or gas
  • DescriptionPaddle Flow Indicator, 0.5 to 10 L/min for 1/4" to 5/16" ID Tubing
Key Features
  • Visually confirm flow with clear window
More About this Item
Indicators give visual confirmation of flow through a clear polystyrene housing. All models can be used with noncorrosive liquids and gases. Connect tubing to indicators via hose barb connections; model 06297-05 has straight ends. All models are pressure rated to 10 psig.