Benchmark Scientific D1032-60 Prefilled 2.0 mL Tubes, Ceria Stabilized Zirconium Satellite, 6 mm; 50/pk
Representative image only.
Benchmark Scientific Mfr # D1032-60 – Item # EW-36271-88
Compatible with all common bead homogenizers
PRODUCT OPTIONS
$185.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • MaterialZirconium
  • DescriptionPrefilled 2.0 mL Tubes, Ceria Stabilized Zirconium Satellite, 6 mm; 50/pk
More About this Item
Glass, zirconium, and stainless steel homogenizer beads, available in a variety of sizes, are efficient and quick at breaking down a wide range of samples in order to release the DNA, RNA, and proteins contained within cells. The beads come in prefilled unskirted tubes or in bulk, and are compatible with all popular bead homogenizers.
Ceria stabilized zirconium beads are quality tested to ensure the elimination of DNase, RNase, protease, and nucleic acids.