Bio Plas Capsette 6036 Cassettes, Burnt orange, 500/pack
Representative image only.
Bio Plas Mfr # 6036 – Item # EW-48620-06
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Capsette cassettes accept standard stainless steel lids
SPECS & DESCRIPTION
Specifications & Description
  • ColorOrange
  • DescriptionCassettes, Burnt orange, 500/pack
Key Features
  • Fit standard stainless steel and disposable plastic base molds
  • Strategically designed slots for efficient solvent flow and drainage
  • Float and remain closed throughout processing
  • Rigid construction eliminates "chattering" during sectioning
  • Acetal copolymer plastic fabrication resists heat, solvents, decalcifying solutions, and effects of microwave processing
  • Accepted by all current automatic cassette labelers
  • Capsette cassettes accept standard stainless steel lids