Bio-Simplex DCB40L-3EZ-ST EZ Top Closure Assembly, Sani-Tech, 3-Port for 40L Carboy
Representative image only.
Bio-Simplex Mfr # DCB40L-3EZ-ST – Item # EW-62500-57
Optimize performance of your Bio-Simplex Polypropylene Carboys
  • Compatible with the Bio-Simplex Carboys
  • Constructed of durable polypropylene
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPolypropylene (PP)
  • Cap / Stopper Size120 mm
  • Cap ColorWhite
  • AutoclavableYes
  • Tubing ID9.5 mm
  • DescriptionEZ Top Closure Assembly, Sani-Tech, 3-Port for 40L Carboy
Key Features
  • Compatible with the Bio-Simplex Carboys
  • Constructed of durable polypropylene
More About this Item
Choose from a wide range of accessories and replacement parts for your Bio-Simplex carboys.
Pre-assembled caps are designed for single use and to withstand temperatures and sterilization to 275°F (135°C). Autoclavable.
Can successfully replace stainless steel closures and eliminate costs for cleaning, validation and assembly. One-piece construction eliminates product loss and contamination due to loose connection. Tubing's smooth inner bore eliminates risk of product entrapment and build-up.
Sani-Tech high-purity, platinum-cured silicone closure has 3 ports and includes an overmolded sinker.