Bio-Works BabyBio Dsalt Pre-Packed Desalting and Buffer Exchange Column; 10 x 1 mL
Representative image only.
Bio-Works Mfr # KN45360104 – Item # EW-11947-73
Designed for rapid and efficient desalting and buffer exchange applications
  • Easy and convenient group separation of high and low molecular weight substances
  • Reproducible and easy-to-use prepacked columns
  • Applications: Targets proteins and other biomolecules of similar size
SPECS & DESCRIPTION
Specifications & Description
  • Max Molecular Weight Cutoff (kDa)5
  • Column Volume (mL/cm)1
  • ApplicationsTargets proteins and other biomolecules of similar size
  • DescriptionPre-Packed Desalting and Buffer Exchange Column; 10 x 1 mL
Key Features
  • Easy and convenient group separation of high and low molecular weight substances
  • Reproducible and easy-to-use prepacked columns
  • Applications: Targets proteins and other biomolecules of similar size
More About this Item
Use size exclusion method to quickly and efficiently separate high molecular weight substances such as proteins and biomolecules from salts and other small molecules. Buffer exchange or desalting of a sample can be used to prepare for mass spectroscopy analysis or lyophilization, and after certain procedures such as ion exchange chromatography. Useful alternative to dialysis for larger or degradable samples. The molecular weight cutoff is 5000. Recommended flow rate is 1 mL/min for 1-mL column, 5 mL/min for 5-mL column. Stable over pH 2 to 12 working range.
Columns feature biocompatible polypropylene body and polyethylene top and bottom filters. Each column comes with a plug in the inlet and a snap-off end at the outlet. Easily connect to a syringe, pump, or chromatography system using fingertight fittings (coned 10-32) for 1/16" OD tubing. Connect up to five columns in series for larger sample volumes.