Bio-Works BabyBio S Pre-Packed Chromatography Column, Sulfonate; 10 x 5 mL
Representative image only.
Bio-Works Mfr # KN45200108 – Item # EW-11947-69
Ready to use for quick and easy purification of proteins
  • Rapid method screening and separation
  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • Applications: Strong cation exchange, 3 to 13 pH
SPECS & DESCRIPTION
Specifications & Description
  • Column Volume (mL/cm)5
  • Wetted MaterialsSulfonate resin
  • Total Volume Capacity (mL)Dynamic binding: 130 mg BSA/mL medium
  • ApplicationsStrong cation exchange, 3 to 13 pH
  • DescriptionPre-Packed Chromatography Column, Sulfonate; 10 x 5 mL
Key Features
  • Rapid method screening and separation
  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • Applications: Strong cation exchange, 3 to 13 pH
More About this Item
BabyBio IEC columns come prepacked with WorkBeads™ media, agarose-based chromatographic resins, which provide a tight size distribution and exceptional mechanical stability. Beads in columns maintain high capacity over broad pH ranges. Connect up to five columns in series to process larger sample volumes.
Columns feature biocompatible polypropylene body and polyethylene top and bottom filters. Each column comes with a plug in the inlet and a snap-off end at the outlet. Easily connect to a syringe, pump, or chromatography system using fingertight fittings (coned 10-32) for 1/16" OD tubing.