Bio-Works WorkBeads 40/10 000 ACT Activated Medium, Bromohydrin, 10M kDa; 1 L
Representative image only.
Bio-Works Mfr # KN40450010 – Item # EW-11947-07
Separate proteins using reverse interaction between the target protein and ligand of bead
  • Reliable, reproducible, and efficient
  • High chemical stability for easy clean-in-place (CIP)
  • Applications: Small molecules, peptides, proteins, immunoglobulins
SPECS & DESCRIPTION
Specifications & Description
  • Media Volume (mL)1000
  • Max Molecular Weight Cutoff (kDa)10M
  • Wetted MaterialsActivated media
  • ApplicationsSmall molecules, peptides, proteins, immunoglobulins
  • DescriptionActivated Medium, Bromohydrin, 10M kDa; 1 L
Key Features
  • Reliable, reproducible, and efficient
  • High chemical stability for easy clean-in-place (CIP)
  • Applications: Small molecules, peptides, proteins, immunoglobulins
More About this Item
Use affinity chromatography to separate biochemical mixtures based on a highly specific interaction between antigen and antibody, enzyme and substrate, or receptor and ligand.
Activated media are a reliable starting material for the preparation of customized chromatography resins. Activated media have a bromohydrin group that is reactive towards primary amines, thiol, hydroxyl, or histidyl residues. Select from two pore sizes depending on your application needs. Use the higher porosity media if the ligand or target is large.
Order media for antibodies for purification of monoclonal and polyclonal antibodies. The specificity of the recombinant Protein A for the Fc-region of IgG provides excellent purification. The Protein A ligand is coupled to the bead using a bromohydrin-based method that gives high chemical stability and low ligand leakage.