Bio-Works WorkBeads 40 DEAE Ion Exchange Medium, Diethylaminoethyl; 25 mL

Representative image only.
Bio-Works Mfr # KN40150001 – Item # EW-11946-87
High-throughput separation of proteins, peptides, and oligonucleotides
  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • High chemical stability for easy clean-in-place (CIP)
  • Efficient separation due to the narrow particle size distribution
  • Applications: Weak anion exchange

SPECS & DESCRIPTION

Specifications & Description

  • Media Volume (mL)25
  • Wetted MaterialsDiethylaminoethyl resin
  • ApplicationsWeak anion exchange
  • DescriptionIon Exchange Medium, Diethylaminoethyl; 25 mL

Key Features

  • Agarose-based resin provides optimal selectivity, resolution, and purity
  • High chemical stability for easy clean-in-place (CIP)
  • Efficient separation due to the narrow particle size distribution
  • Applications: Weak anion exchange

More About this Item

Manufactured using agarose-based chromatographic resins, WorkBeads ion exchange purification media provide a tight size distribution and exceptional mechanical stability. Average bead size of 40 µm enables high-resolution separation at flows up to 700 cm/hr at moderate backpressure. Stable over pH 2 to 13 range.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat