Bio-Works WorkBeads 40 SEC Size Exclusion Medium, 50 to 1200 kDa; 300 mL

Representative image only.
Bio-Works Mfr # KN40300003 – Item # EW-11946-94
Separate molecules based on their size by filtration through the gel
  • Great selectivity and efficiency results in excellent resolution
  • High chemical stability for easy clean-in-place (CIP)
  • Applications: Purification

SPECS & DESCRIPTION

Specifications & Description

  • Media Volume (mL)300
  • Min Molecular Weight Cutoff (kDa)50
  • Max Molecular Weight Cutoff (kDa)1200
  • ApplicationsPurification
  • DescriptionSize Exclusion Medium, 50 to 1200 kDa; 300 mL

Key Features

  • Great selectivity and efficiency results in excellent resolution
  • High chemical stability for easy clean-in-place (CIP)
  • Applications: Purification

More About this Item

Separation by size exclusion occurs when different size molecules are included or excluded from the pores of the matrix. Larger molecules, too big to enter the pores, flow quickly down the column while smaller-sized molecules enter the pores and pass through, resulting in slower progress through the media. This method is great for separating large molecules from the small molecules while preserving the biological activity of the particles to be separated. Recommended flow rate of 600 cm/hr. Stable over pH 2 to 13 working range.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat