BioTek ELX800 Microplate reader, 400 to 750 nm, 6, 12, 24, 48, 96 well

Representative image only.
BioTek Mfr # ELX800 – Item # EW-13055-50
Optional 72/96 Terasaki and 3840well capacity, UV 340nm capability
  • Stores up to 75 custom assay definitions and 8 test results
  • Extensive onboard data analysis and optional software which offers more flexible data manipulation
  • 1 year warranty

SPECS & DESCRIPTION

Specifications & Description

  • Min Wavelength (nm)400
  • Max Wavelength (nm)750
  • Light Sourcetungsten halogen bulb
  • Linearity1% from 0 to 2.0 OD
  • Height (in)16 1/2
  • Width (in)7
  • Length (in)15
  • Height (mm)419
  • Width (mm)178
  • Length (mm)384
  • Power (VAC)100 to 240
  • Power (Hz)50/60
  • DescriptionMicroplate reader, 400 to 750 nm, 6, 12, 24, 48, 96 well
  • Warranty1 year
1 Year WarrantyETL Listed

Key Features

  • Stores up to 75 custom assay definitions and 8 test results
  • Extensive onboard data analysis and optional software which offers more flexible data manipulation
  • 1 year warranty

More About this Item

The Elx800 microplate readers feature on board data reduction and reporting. Reads 6-, 12-, 24--, 48-, and 96-well plate microplates. The model 13055-52 can read 384-well and 60-, 72- and 96-well Terasaki plates. Wavelength range from 400 to 750nm. Model 13055-54 optics to extend wavelength range down to 340nm.

Microplate readers feature a 2x24-character LCD and convenient soft-keys. Extensive curve fitting options include linear, cubic, quadratic, 4-p, log-logit, cubicspline, and point-to-point. Memory stores 25 standard curves and up to 8 microplate test results. Features auto or manual plate layout for blanks, controls, standards, and samples. Also offers control and assay validation, transformation and formula applications, and cutoff and call criteria.

Onboard software is easy to program to run sophisticated assays and analyzed without the need of computers. Additional software 13055-56 offers easy user interface with flexible data manipulation and curve fitting for basic endpoint, kinetic, multi-wavelength and spectral scan requirements. Software package includes software on CD, and three user licenses. What’s included: 340 (13055-54 only), 405, 450, 490, 630nm filters, dust cover, parallel printer cable (25-pin male and 36-pin male), serial cable (null modem, 9-pin female to 25-pin female), operator’s manual. Additional filters are available range from 304 to 750nm.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat