Bradley S19214PDC Eyewash, Pedestal mount, ABS plastic with cover
Representative image only.
Bradley Mfr # S19214PDC – Item # EW-86001-26
No struggling to activate the eyewash – broad hand flag makes activation easy!
  • Eye wash bowls provide excellent face wash down coverage
PRODUCT OPTIONS
$448.00USD / Each
SPECS & DESCRIPTION
Specifications & Description
  • Eyewash Fixture MaterialABS plastic
  • Bowl MaterialABS plastic
  • MountPedestal Mount
  • Outlet Connections1-1/4" NPT(M)
  • Inlet Connection1/2" NPT(F)
  • ComplianceANSI Z358.1-2009
  • DescriptionEyewash, Pedestal mount, ABS plastic with cover
Includes
universal eyewash sign and inspection tag.
Key Features
  • Eye wash bowls provide excellent face wash down coverage
More About this Item
HALO eyewashes provide rapid relief to individuals who have been injured by chemicals or particulates. Ergonomic hand activation is easy to reach from both sides of the bowl. Integral strainer reduces debris in the supply water for further eye protection. Bowls are contoured for superior draining, reducing the risk of bacterial growth when not in use. Separate supply and waste pipes meet CEN and plumbing requirements. Supply connection is 1/2" NPT(F); waste connection is 1-1/4" NPT(M).
Compliance: ANSI Z358.1-2009
ABS plastic models with covers feature a translucent plastic cover for easy inspection.