Brady M21-375-580-WT BMP 21 Label Cartridge; 3/8"W, Permanent Vinyl, White
Representative image only.
Brady Mfr # M21-375-580-WT – Item # EW-65533-16
Identify your lab samples quickly and legibly
  • One-handed operation with large display and backlight
  • Advanced serialization, barcoding and symbols
  • Cuts and holds printed labels until you need them
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionBMP 21 Label Cartridge; 3/8\"W, Permanent Vinyl, White
Key Features
  • One-handed operation with large display and backlight
  • Advanced serialization, barcoding and symbols
  • Cuts and holds printed labels until you need them
More About this Item
The BMP 21 portable label printer is an all-in-one labeler. With label materials designed for a variety of environments, the BMP 21 portable printer can quickly and easily create clear, legible labels that stay in place for years — despite extreme temperatures and curved or highly textured surfaces. Printer operates on either six AA batteries or AC power adapter (neither included; order separately).
Print labels in sizes specifically made for vials and slides. Vial size fonts are preprogrammed for 0.6, 1.5, and 2.0 mL. The laboratory-specific labels resist most chemicals, including alcohol, ethanol, and the solvent DMSO. Choose from 3/8", 1/2", and 3/4" wide labels in white or clear tape. Labels are designed for normal to extreme conditions, like liquid nitrogen or hot water baths.