BrandTech 115016 Sample Cup for Technicon® Analyzers, PS, 2 mL; 10,000/PK

Representative image only.
BrandTech Mfr # 115016 – Item # EW-06101-35
Designed to fit common analyzers
  • Clear polystyrene sample cups provide quick and acurate analysis
  • For Technicon® analyzers
  • 2 mL capacity
  • 14.8 mm top dia x 12.8 mm bottom dia

$1,106.00USD / Pkg of 10000

SPECS & DESCRIPTION

Specifications & Description

  • Capacity (mL)2
  • MaterialPolystyrene
  • ColorClear
  • Compatible WithTechnicon® analyzers
  • Height (mm)24.9
  • DescriptionSample Cup for Technicon® Analyzers, PS, 2 mL; 10,000/PK

Key Features

  • Clear polystyrene sample cups provide quick and acurate analysis
  • For Technicon® analyzers
  • 2 mL capacity
  • 14.8 mm top dia x 12.8 mm bottom dia

More About this Item

Order push-on caps to securely contain your sample.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat