BrandTech 701358 Sealing Mat for Deep Well Plate, 96-Well, PP, 0.5 mL, Standard; 50/PK
Representative image only.
BrandTech Mfr # 701358 – Item # EW-07904-35
Provide effective plug seal for each well
  • Reduces evaporation of sample
  • Improves sealing
  • Sealing Mat for Standard 96-Well, Deep Well Plate Capacity: 0.5 mL Made of Polypropylene
PRODUCT OPTIONS
$419.00USD / Pkg of 50
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)0.5
  • MaterialPolypropylene
  • DescriptionBrandTech Sealing Mat for Deep Well Plate, 96-Well, PP, 0.5 mL, Standard; 50/PK
Key Features
  • Reduces evaporation of sample
  • Improves sealing
  • Sealing Mat for Standard 96-Well, Deep Well Plate Capacity: 0.5 mL Made of Polypropylene
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. The mats are made of silicone, polypropylene, or modified polyethylene.