BrandTech Reagent Reservoirs, Pyramid Bottom, Polypropylene, 6 Columns, 40 mL per Column; 10/Pk
Representative image only.
BrandTech Mfr # 701456 – Item # EW-06039-04
Ideal for repetitive liquid transfer with pipettors
  • Constructed of durable polypropylene
  • Translucent for optimal solution visibility
  • Made of translucent polyproplyene
  • 6 columns with 40 mL volume per column
SPECS & DESCRIPTION
Specifications & Description
  • SterileNo
  • Max Volume (mL)240
  • Graduation Interval40 mL per Column
  • Number Of Channels6
  • Wetted MatterialPolypropylene
  • DescriptionReagent Reservoirs, Pyramid Bottom, Polypropylene, 6 Columns, 40 mL per Column; 10/Pk
Key Features
  • Constructed of durable polypropylene
  • Translucent for optimal solution visibility
  • Made of translucent polyproplyene
  • 6 columns with 40 mL volume per column
More About this Item
Single-use reservoirs save you the hassle of washing and the problem of cross-contamination.