BRAND Transferpettor 702854 Positive Displacement Pipettor Tips, PP, 200-1000 µL, 10/PK

Representative image only.
Mfr # 702854 – Item # EW-24550-76
Color coded to match appropriate sized pipettes
  • Robust polypropylene (PP) construction
  • Reusable

PRODUCT OPTIONS

$39.50USD / Pkg of 10

SPECS & DESCRIPTION

Specifications & Description

  • SterileNo
  • Max Volume (µL)1000
  • Min Volume (µL)200
  • Min Volume (mL)0.2
  • Max Volume (mL)1
  • Tip Volume (mL)1
  • Min Dispensing (mL)0.2
  • Max Dispensing (mL)1
  • Wetted MatterialPolypropylene
  • Color CodeYellow
  • DescriptionPositive Displacement Pipettor Tips, PP, 200-1000 µL, 10/pk

Key Features

  • Robust polypropylene (PP) construction
  • Reusable

More About this Item

Reusable tips are specially designed for the Transferpettor positive displacement pipettor.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At1-800-323-4340
Live chat