BRAND 704322 Threaded Adapter for Dispensers and Digital Burettes, PP, 22 mm x 33 mm; 1/EA
Representative image only.
Mfr # 704322 – Item # EW-07910-77
Accessories to fit Dispensette® and seripettor® bottletop dispensers and Titrette® bottletop burettes
  • Maximize safety, convenience and productivity
  • Easily install drying tube to protect against humidity and carbon dioxide
  • Simple serial dispensing with flexible discharge tube kits
  • Replacement parts to extend the life of your instrument
  • Stacks with 704395 to fit Dispensette®, Titrette® and seripettor® (all models)
SPECS & DESCRIPTION
Specifications & Description
  • Description704322 Threaded Adapter for Dispensers and Digital Burettes, PP, 22 mm x 33 mm; 1/EA
Key Features
  • Maximize safety, convenience and productivity
  • Easily install drying tube to protect against humidity and carbon dioxide
  • Simple serial dispensing with flexible discharge tube kits
  • Replacement parts to extend the life of your instrument
  • Stacks with 704395 to fit Dispensette®, Titrette® and seripettor® (all models)
More About this Item
These accessories consist of durable parts that come in handy to enhance the capabilities of bottletop dispensers or burettes.
An optional drying tube that screws into the accessory port of a Dispensette® or Titrette® and protects sensitive reagents from humidity and carbon dioxide is among a wide range of accessories. There also are Flexible Discharge Tube Kits for Dispensette® S Dispensers for serial dispensing. The kits contain a flexible discharge tube with a textured safety handle for quick and accurate dispensing, and valve to help rid away with bubbles (not for use with HF).
Replacement parts includes filling valves and inspection windows for the Titrette®. There is the Safety Prime™ Venting Valves to be used with the Dispensette® Organic and Dispensette® III Bottletop Dispensers. The valves come complete with a seal and recirculation tube.
A selection of polypropylene and fluoropolymer thread adapters, and burette reservoir bottles are also listed.