BrandTech 705748 PD-Tip™ BIO-CERT® Tip, Sterile, 50 mL; 25/pk
Representative image only.
BrandTech Mfr # 705748 – Item # EW-24806-27
The perfect tips for your repeating pipette
  • High quality virgin plastic construction
  • Encoded for automatic recognition
  • DNA, RNAse, endotoxin, and ATP free
  • Available sterile and non-sterile
PRODUCT OPTIONS
$48.00USD / Pkg of 25
SPECS & DESCRIPTION
Specifications & Description
  • SterileYes
  • Tip Volume (mL)50
  • Wetted MatterialPolyethylene and polypropylene
  • Filter TypeNone
  • Package Of25
  • DescriptionPD-Tip™ BIO-CERT® Sterile, 50 mL; 25/pk
Key Features
  • High quality virgin plastic construction
  • Encoded for automatic recognition
  • DNA, RNAse, endotoxin, and ATP free
  • Available sterile and non-sterile
More About this Item
PD-Tip™ Precision Dispenser tips are constructed of high-quality virgin polyethylene and polypropylene, except for the 0.1-mL tip which is polyethylene. An extended piston to minimize dead volume and maximize fluid utilization. The precision seal between the piston and cylinder ensures smooth operation, increased accuracy, and increased precision while pipetting. Sterile tips are individually wrapped and certified free of DNA, RNAse, endotoxins, and ATP. 25 and 50 mL tip packs include one autoclavable, polypropylene adapter.